2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Bromo-6-chlorophenyl)-2,2,2-trifluoroethanone
PC502434-1G
PC502434-25G
PC502434-5G
APOSPC502434-1GEA
45.1
EUR
APOSPC502434-1G
APOSPC502434-25G
APOSPC502434-5G
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone 96%
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone
Formel:
C₈H₃BrClF₃O Molecular Weight: 287,461 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062425 CAS-Nummer: 886502-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...