3-Bromo-4-fluoro-2-nitrophenol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502440-1G
PC502440-250MG
APOSPC502440-1GEA
609
EUR
APOSPC502440-1G
APOSPC502440-250MG
3-Bromo-4-fluoro-2-nitrophenol 95%
3-Bromo-4-fluoro-2-nitrophenol
Formel:
C₆H₃BrFNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23132693 CAS-Nummer: 1374772-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...