1,3,5-Tribromo-2,4,6-trifluorobenzene 97%
Lieferant: Apollo Scientific
PC502442-1G
PC502442-250MG
APOSPC502442-1GEA
98.6
EUR
APOSPC502442-1G
APOSPC502442-250MG
1,3,5-Tribromo-2,4,6-trifluorobenzene 97%
1,3,5-Tribromo-2,4,6-trifluorobenzene
Formel:
C₆Br₃F₃ Molecular Weight: 368.771 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192722 CAS-Nummer: 2368-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...