3,6-Difluoro-2-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC502477-1G PC502477-250MG PC502477-5G
APOSPC502477-1GEA 58 EUR
APOSPC502477-1G APOSPC502477-250MG APOSPC502477-5G
3,6-Difluoro-2-(trifluoromethoxy)benzyl alcohol 95%
3,6-Difluoro-2-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₅F₅O₂
Molecular Weight: 228,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561160
CAS-Nummer: 2244081-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR