2,6-Difluoro-4-(trifluoromethoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC502502-1G PC502502-250MG PC502502-5G
APOSPC502502-1GEA 0 EUR
APOSPC502502-1G APOSPC502502-250MG APOSPC502502-5G
2,6-Difluoro-4-(trifluoromethoxy)benzoic acid 95%
2,6-Difluoro-4-(trifluoromethoxy)benzoic acid
Formel: C₈H₃F₅O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31561167
CAS-Nummer: 2244079-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR