2,6-Difluoro-4-(trifluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC502511-1G PC502511-250MG PC502511-5G
APOSPC502511-1GEA 30.6 EUR
APOSPC502511-1G APOSPC502511-250MG APOSPC502511-5G
2,6-Difluoro-4-(trifluoromethoxy)benzyl bromide 95%
2,6-Difluoro-4-(trifluoromethoxy)benzyl bromide
Formel: C₈H₄BrF₅O
Molecular Weight: 291.013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561169
CAS-Nummer: 2244082-19-5
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR