2,3-Difluoro-4-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC502528-1G PC502528-250MG PC502528-5G
APOSPC502528-1GEA 58 EUR
APOSPC502528-1G APOSPC502528-250MG APOSPC502528-5G
2,3-Difluoro-4-(trifluoromethoxy)benzyl alcohol 95%
2,3-Difluoro-4-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₅F₅O₂
Molecular Weight: 228.116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561152
CAS-Nummer: 2244080-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR