3-Fluoro-2-iodobenzonitrile

Lieferant: Apollo Scientific

PC502585-5G
APOSPC502585-5GEA 184 EUR
APOSPC502585-5G
3-Fluoro-2-iodobenzonitrile
3-Fluoro-2-iodobenzonitrile
Formel: C₇H₃FIN
Molecular Weight: 247,008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782075
CAS-Nummer: 916792-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR