2-Iodo-5-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific

PC502631-1G PC502631-5G
APOSPC502631-1GEA 102 EUR
APOSPC502631-1G APOSPC502631-5G
2-Iodo-5-(trifluoromethyl)benzonitrile
2-Iodo-5-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₃IN
Molecular Weight: 297.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398159
CAS-Nummer: 868166-20-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR