3-Nitro-5-(trifluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502653-10G
PC502653-1G
PC502653-5G
PC502653-25G
APOSPC502653-10GEA
1020
EUR
APOSPC502653-10G
APOSPC502653-1G
APOSPC502653-5G
APOSPC502653-25G
3-Nitro-5-(trifluoromethyl)benzaldehyde 95%
3-Nitro-5-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄F₃NO₃ Molecular Weight: 219.117 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847795 CAS-Nummer: 284047-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...