[3-Nitro-5-(trifluoromethyl)phenyl]methanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Nitro-5-(trifluoromethyl)phenyl)methanol
PC502654-1G
PC502654-250MG
PC502654-5G
APOSPC502654-1GEA
69.6
EUR
APOSPC502654-1G
APOSPC502654-250MG
APOSPC502654-5G
[3-Nitro-5-(trifluoromethyl)phenyl]methanol 95%
[3-Nitro-5-(trifluoromethyl)phenyl]methanol
Formel:
C₈H₆F₃NO₃ Molecular Weight: 221.133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040241 CAS-Nummer: 180146-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...