1-(Bromomethyl)-3-nitro-5-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific
Gefahr

PC502655-1G PC502655-250MG PC502655-5G
APOSPC502655-1GEA 296 EUR
APOSPC502655-1G APOSPC502655-250MG APOSPC502655-5G
1-(Bromomethyl)-3-nitro-5-(trifluoromethyl)benzene 95%
1-(Bromomethyl)-3-nitro-5-(trifluoromethyl)benzene
Formel: C₈H₅BrF₃NO₂
Molecular Weight: 284.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044988
CAS-Nummer: 180146-67-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR