4-Nitro-3-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific

PC502660-1G PC502660-250MG PC502660-5G
APOSPC502660-1GEA 136 EUR
APOSPC502660-1G APOSPC502660-250MG APOSPC502660-5G
4-Nitro-3-(trifluoromethyl)benzaldehyde
4-Nitro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₃NO₃
Molecular Weight: 219.117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185770
CAS-Nummer: 101066-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR