2-Chloro-6-(trifluoromethoxy)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502715-1G
PC502715-250MG
PC502715-100MG
PC502715-5G
APOSPC502715-1GEA
290
EUR
APOSPC502715-1G
APOSPC502715-250MG
APOSPC502715-100MG
APOSPC502715-5G
2-Chloro-6-(trifluoromethoxy)benzaldehyde 95%
2-Chloro-6-(trifluoromethoxy)benzaldehyde
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224,564 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393770 CAS-Nummer: 1261822-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...