2-Chloro-6-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC502716-1G PC502716-250MG
APOSPC502716-1GEA 723 EUR
APOSPC502716-1G APOSPC502716-250MG
2-Chloro-6-(trifluoromethoxy)benzyl alcohol 95%
2-Chloro-6-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆ClF₃O₂
Molecular Weight: 226.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393817
CAS-Nummer: 1261791-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR