Ethyl 2-amino-4-(difluoromethyl)-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

PC50280-1G PC50280-250MG
APOSPC50280-1GEA 618 EUR
APOSPC50280-1G APOSPC50280-250MG
Ethyl 2-amino-4-(difluoromethyl)-1,3-thiazole-5-carboxylate
Ethyl 2-amino-4-(difluoromethyl)-1,3-thiazole-5-carboxylate
Formel: C₇H₈F₂N₂O₂S
Molecular Weight: 222,212 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30471216
CAS-Nummer: 1631960-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR