4-Bromo-3-iodobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC502881-250MG
APOSPC502881-250MGEA 79.9 EUR
APOSPC502881-250MG
4-Bromo-3-iodobenzyl fluoride 95%
4-Bromo-3-iodobenzyl fluoride
Formel: C₇H₅BrFI
Molecular Weight: 314,921 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31634827
CAS-Nummer: 2244083-96-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR