2-Bromo-6-iodobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC502883-1G
PC502883-250MG
APOSPC502883-1GEA
609
EUR
APOSPC502883-1G
APOSPC502883-250MG
2-Bromo-6-iodobenzyl fluoride 95%
2-Bromo-6-iodobenzyl fluoride
Formel:
C₇H₅BrFI Molecular Weight: 314,921 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31634828 CAS-Nummer: 2244087-75-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...