1,1,1,3,3,3-Hexafluoro-2-(3-nitrophenyl)propan-2-ol 95%

Lieferant: Apollo Scientific
Achtung

PC50362-1G PC50362-250MG
APOSPC50362-1GEA 574 EUR
APOSPC50362-1G APOSPC50362-250MG
1,1,1,3,3,3-Hexafluoro-2-(3-nitrophenyl)propan-2-ol 95%
1,1,1,3,3,3-Hexafluoro-2-(3-nitrophenyl)propan-2-ol
Formel: C₉H₅F₆NO₃
Molecular Weight: 289,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123441
CAS-Nummer: 2402-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR