2-Bromo-1-fluoro-5-methyl-3-nitrobenzene 95%

Lieferant: Apollo Scientific
Achtung

PC50363-1G PC50363-250MG PC50363-5G
APOSPC50363-1GEA 37.7 EUR
APOSPC50363-1G APOSPC50363-250MG APOSPC50363-5G
2-Bromo-1-fluoro-5-methyl-3-nitrobenzene 95%
2-Bromo-1-fluoro-5-methyl-3-nitrobenzene
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28740570
CAS-Nummer: 1807212-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR