2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95+%

Lieferant: Apollo Scientific
Achtung

PC50370-1G PC50370-250MG PC50370-5G
APOSPC50370-1GEA 300 EUR
APOSPC50370-1G APOSPC50370-250MG APOSPC50370-5G
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95+%
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formel: C₉H₇F₆NO
Molecular Weight: 259.148 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17015636
CAS-Nummer: 2402-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR