5-Fluoro-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluorobenzofuran-2-carboxylic acid

PC50380-1G PC50380-250MG PC50380-5G
APOSPC50380-1GEA 66.7 EUR
APOSPC50380-1G APOSPC50380-250MG APOSPC50380-5G
5-Fluoro-1-benzofuran-2-carboxylic acid
5-Fluoro-1-benzofuran-2-carboxylic acid
Formel: C₉H₅FO₃
Molecular Weight: 180,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038172
CAS-Nummer: 89197-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR