6-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC50387-100MG
PC50387-250MG
APOSPC50387-100MGEA
139
EUR
APOSPC50387-100MG
APOSPC50387-250MG
6-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
6-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Formel:
C₁₀H₅FO₄ Molecular Weight: 208.143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08753858 CAS-Nummer: 937688-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...