4'-Chloro-3'-fluorostyrene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-3-fluorostyrene
PC50392-1G
PC50392-250MG
PC50392-5G
APOSPC50392-1GEA
60.9
EUR
APOSPC50392-1G
APOSPC50392-250MG
APOSPC50392-5G
4'-Chloro-3'-fluorostyrene
4'-Chloro-3'-fluorostyrene
Formel:
C₈H₆ClF Molecular Weight: 156,585 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20384550 CAS-Nummer: 1263414-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...