2-Fluoro-6-hydrazinopyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Fluoropyridin-2-yl)hydrazine

PC50396-1G PC50396-250MG PC50396-5G
APOSPC50396-1GEA 46.4 EUR
APOSPC50396-1G APOSPC50396-250MG APOSPC50396-5G
2-Fluoro-6-hydrazinopyridine 97%
2-Fluoro-6-hydrazinopyridine
Formel: C₅H₆FN₃
Molecular Weight: 127.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214745
CAS-Nummer: 80714-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR