3',4'-Difluoro[1,1'-biphenyl]-4-amine 95%

Lieferant: Apollo Scientific
Achtung

PC50398-1G PC50398-250MG
APOSPC50398-1GEA 214 EUR
APOSPC50398-1G APOSPC50398-250MG
3',4'-Difluoro[1,1'-biphenyl]-4-amine 95%
3',4'-Difluoro[1,1'-biphenyl]-4-amine
Formel: C₁₂H₉F₂N
Molecular Weight: 205,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12779907
CAS-Nummer: 1184136-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR