1-(3-Bromoprop-1-ynyl)-3-fluoro-benzene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Bromoprop-1-yn-1-yl)-3-fluorobenzene

PC50402-1G PC50402-250MG PC50402-5G
APOSPC50402-1GEA 84.1 EUR
APOSPC50402-1G APOSPC50402-250MG APOSPC50402-5G
1-(3-Bromoprop-1-ynyl)-3-fluoro-benzene 98%
1-(3-Bromoprop-1-ynyl)-3-fluoro-benzene
Formel: C₉H₆BrF
Molecular Weight: 213,046 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD22482456
CAS-Nummer: 1020153-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR