3,4-Dichloro-5-(trifluoromethyl)bromobenzene 95%
Lieferant: Apollo Scientific
PC50417-10G
PC50417-1G
PC50417-5G
APOSPC50417-10GEA
632
EUR
APOSPC50417-10G
APOSPC50417-1G
APOSPC50417-5G
3,4-Dichloro-5-(trifluoromethyl)bromobenzene 95%
3,4-Dichloro-5-(trifluoromethyl)bromobenzene
Formel:
C₇H₂BrCl₂F₃ Molecular Weight: 293,896 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226209 CAS-Nummer: 392-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...