3,5-Dichloro(pentafluorosulphur)benzene 98%
Lieferant: Apollo Scientific
PC50418-10G
PC50418-1G
PC50418-5G
APOSPC50418-10GEA
1900
EUR
APOSPC50418-10G
APOSPC50418-1G
APOSPC50418-5G
3,5-Dichloro(pentafluorosulphur)benzene 98%
3,5-Dichloro(pentafluorosulphur)benzene
Formel:
C₆H₃Cl₂F₅S Molecular Weight: 273.051 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32708471 CAS-Nummer: 159727-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...