1-Butoxy-3,5-difluorobenzene 98%
Lieferant: Apollo Scientific
PC50423-1G
PC50423-250MG
PC50423-5G
APOSPC50423-1GEA
369
EUR
APOSPC50423-1G
APOSPC50423-250MG
APOSPC50423-5G
1-Butoxy-3,5-difluorobenzene 98%
1-Butoxy-3,5-difluorobenzene
Formel:
C₁₀H₁₂F₂O Molecular Weight: 186,198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13250188 CAS-Nummer: 123843-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...