2,4-Difluorophenetole 98%

Lieferant: Apollo Scientific

Synonyme: 1-Ethoxy-2,4-difluorobenzene

PC50425-10G PC50425-1G PC50425-5G
APOSPC50425-10GEA 574 EUR
APOSPC50425-10G APOSPC50425-1G APOSPC50425-5G
2,4-Difluorophenetole 98%
2,4-Difluorophenetole
Formel: C₈H₈F₂O
Molecular Weight: 158.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846411
CAS-Nummer: 348-20-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR