3,5-Difluorophenetole 98%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-5-ethoxybenzene
PC50429-10G
PC50429-1G
PC50429-5G
APOSPC50429-10GEA
480
EUR
APOSPC50429-10G
APOSPC50429-1G
APOSPC50429-5G
3,5-Difluorophenetole 98%
3,5-Difluorophenetole
Formel:
C₈H₈F₂O Molecular Weight: 158,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19105371 CAS-Nummer: 144891-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...