3-Fluoro-5-(pentafluorosulphur)bromobenzene 98%
Lieferant: Apollo Scientific
PC50430-1G
PC50430-5G
APOSPC50430-1GEA
696
EUR
APOSPC50430-1G
APOSPC50430-5G
3-Fluoro-5-(pentafluorosulphur)bromobenzene 98%
3-Fluoro-5-(pentafluorosulphur)bromobenzene
Formel:
C₆H₃BrF₆S Molecular Weight: 301,047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32708474 CAS-Nummer: 2514942-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...