4-Fluoro-3-methyl-5-(trifluoromethyl)bromobenzene 97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-fluoro-1-methyl-3-(trifluoromethyl)benzene

PC50433-10G PC50433-1G PC50433-5G
APOSPC50433-10GEA 574 EUR
APOSPC50433-10G APOSPC50433-1G APOSPC50433-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)bromobenzene 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)bromobenzene
Formel: C₈H₅BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD31737212
CAS-Nummer: 2167934-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR