4-Phenoxy-2-(trifluoromethyl)bromobenzene 97%

Lieferant: Apollo Scientific

PC50434-10G PC50434-1G PC50434-5G
APOSPC50434-10GEA 574 EUR
APOSPC50434-10G APOSPC50434-1G APOSPC50434-5G
4-Phenoxy-2-(trifluoromethyl)bromobenzene 97%
4-Phenoxy-2-(trifluoromethyl)bromobenzene
Formel: C₁₃H₈BrF₃O
Molecular Weight: 317.101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28142610
CAS-Nummer: 54846-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR