3-Bromo-2,4,5-trifluoronitrobenzene 95%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-1,2,4-trifluoro-5-nitrobenzene

PC50477-1G PC50477-5G PC50477-10G
APOSPC50477-1GEA 27.7 EUR
APOSPC50477-1G APOSPC50477-5G APOSPC50477-10G
3-Bromo-2,4,5-trifluoronitrobenzene 95%
3-Bromo-2,4,5-trifluoronitrobenzene
Formel: C₆HBrF₃NO₂
Molecular Weight: 255.977 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123591
CAS-Nummer: 485318-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR