2-(Trifluoromethyl)-1H-indol-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
PC50532-1G
PC50532-250MG
PC50532-5G
APOSPC50532-1GEA
125
EUR
APOSPC50532-1G
APOSPC50532-250MG
APOSPC50532-5G
2-(Trifluoromethyl)-1H-indol-4-amine
2-(Trifluoromethyl)-1H-indol-4-amine
Formel:
C₉H₇F₃N₂ Molecular Weight: 200,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28634315 CAS-Nummer: 1553181-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...