2-(Trifluoromethyl)-1H-indol-6-amine

Lieferant: Apollo Scientific
Achtung

PC50533-1G PC50533-250MG PC50533-5G
APOSPC50533-1GEA 574 EUR
APOSPC50533-1G APOSPC50533-250MG APOSPC50533-5G
2-(Trifluoromethyl)-1H-indol-6-amine
2-(Trifluoromethyl)-1H-indol-6-amine
Formel: C₉H₇F₃N₂
Molecular Weight: 200,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20227048
CAS-Nummer: 1236061-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR