Methyl 5-bromo-4-fluorothiophene-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC50535-1G PC50535-250MG
APOSPC50535-1GEA 865 EUR
APOSPC50535-1G APOSPC50535-250MG
Methyl 5-bromo-4-fluorothiophene-2-carboxylate 95%
Methyl 5-bromo-4-fluorothiophene-2-carboxylate
Formel: C₆H₄BrFO₂S
Molecular Weight: 239.062 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10566746
CAS-Nummer: 395664-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR