5-Bromo-4-fluorothiophene-2-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

PC50536-100MG PC50536-1G PC50536-250MG
APOSPC50536-100MGEA 65.4 EUR
APOSPC50536-100MG APOSPC50536-1G APOSPC50536-250MG
5-Bromo-4-fluorothiophene-2-carboxylic acid 96%
5-Bromo-4-fluorothiophene-2-carboxylic acid
Formel: C₅H₂BrFO₂S
Molecular Weight: 225,036 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28975142
CAS-Nummer: 1935274-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR