4-Bromo-2,3,6-trifluorobenzoic acid 95%

Lieferant: Apollo Scientific

PC50545-1G PC50545-250MG PC50545-5G
APOSPC50545-1GEA 284 EUR
APOSPC50545-1G APOSPC50545-250MG APOSPC50545-5G
4-Bromo-2,3,6-trifluorobenzoic acid 95%
4-Bromo-2,3,6-trifluorobenzoic acid
Formel: C₇H₂BrF₃O₂
Molecular Weight: 254,989 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203571
CAS-Nummer: 415965-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR