2-Ethynyl-6-fluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC50559-1G PC50559-250MG
APOSPC50559-1GEA 609 EUR
APOSPC50559-1G APOSPC50559-250MG
2-Ethynyl-6-fluorobenzonitrile 95%
2-Ethynyl-6-fluorobenzonitrile
Formel: C₉H₄FN
Molecular Weight: 145,133 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11656150
CAS-Nummer: 1233509-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR