5-Ethynyl-2-fluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC50565-1G PC50565-250MG
APOSPC50565-1GEA 545 EUR
APOSPC50565-1G APOSPC50565-250MG
5-Ethynyl-2-fluorobenzonitrile 95%
5-Ethynyl-2-fluorobenzonitrile
Formel: C₉H₄FN
Molecular Weight: 145,133 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD14584650
CAS-Nummer: 1093306-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR