2,4-Difluoro-3-(trifluoromethoxy)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC508102-1G PC508102-250MG
APOSPC508102-1GEA 522 EUR
APOSPC508102-1G APOSPC508102-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzyl alcohol
2,4-Difluoro-3-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₅F₅O₂
Molecular Weight: 228.116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723443
CAS-Nummer: 2149598-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR