8-Bromo-5-(trifluoromethyl)quinolin-4(1H)-one

Lieferant: Apollo Scientific
Achtung

PC508114-1G PC508114-250MG
APOSPC508114-1GEA 241 EUR
APOSPC508114-1G APOSPC508114-250MG
8-Bromo-5-(trifluoromethyl)quinolin-4(1H)-one
8-Bromo-5-(trifluoromethyl)quinolin-4(1H)-one
Formel: C₁₀H₅BrF₃NO
Molecular Weight: 292,052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31745736
CAS-Nummer: 1065092-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR