5-Bromo-8-fluoroquinolin-4(1H)-one

Lieferant: Apollo Scientific
Achtung

PC508118-1G PC508118-250MG
APOSPC508118-1GEA 406 EUR
APOSPC508118-1G APOSPC508118-250MG
5-Bromo-8-fluoroquinolin-4(1H)-one
5-Bromo-8-fluoroquinolin-4(1H)-one
Formel: C₉H₅BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD12137226
CAS-Nummer: 1065092-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR