7-Chloro-8-fluoroquinolin-4(1H)-one

Lieferant: Apollo Scientific
Achtung

PC508124-1G PC508124-250MG
APOSPC508124-1GEA 241 EUR
APOSPC508124-1G APOSPC508124-250MG
7-Chloro-8-fluoroquinolin-4(1H)-one
7-Chloro-8-fluoroquinolin-4(1H)-one
Formel: C₉H₅ClFNO
Molecular Weight: 197,594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12192828
CAS-Nummer: 1019016-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR