8-Bromo-4-chloro-6-(trifluoromethyl)quinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC508126-1G
PC508126-250MG
APOSPC508126-1GEA
281
EUR
APOSPC508126-1G
APOSPC508126-250MG
8-Bromo-4-chloro-6-(trifluoromethyl)quinoline
8-Bromo-4-chloro-6-(trifluoromethyl)quinoline
Formel:
C₁₀H₄BrClF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD28010393 CAS-Nummer: 1065093-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...