5-Bromo-4-chloro-8-fluoroquinoline

Lieferant: Apollo Scientific
Achtung

PC508135-1G PC508135-250MG
APOSPC508135-1GEA 281 EUR
APOSPC508135-1G APOSPC508135-250MG
5-Bromo-4-chloro-8-fluoroquinoline
5-Bromo-4-chloro-8-fluoroquinoline
Formel: C₉H₄BrClFN
Molecular Weight: 260,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12137225
CAS-Nummer: 1065093-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR