4,7-Dichloro-8-fluoroquinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC508140-1G
PC508140-250MG
APOSPC508140-1GEA
859
EUR
APOSPC508140-1G
APOSPC508140-250MG
4,7-Dichloro-8-fluoroquinoline
4,7-Dichloro-8-fluoroquinoline
Formel:
C₉H₄Cl₂FN Molecular Weight: 216.039 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09861350 CAS-Nummer: 773148-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...